missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ytterbium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 264300100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Ytterbium(III) oxide | |
99% | |
White | |
10 g | |
O3Yb2 | |
MFCD00011290 | |
Solubility in water: insoluble. Other solubilities: soluble in hot dilute acids | |
[O--].[O--].[O--].[Yb+3].[Yb+3] | |
394.09 | |
99.99% | |
Glass bottle |
2346.0°C | |
1314-37-0 | |
Powder | |
99.99% | |
Yb2O3 | |
14,10105 | |
UZLYXNNZYFBAQO-UHFFFAOYSA-N | |
diytterbium(3+) trioxidandiide | |
394.08 | |
Trace Metal Basis | |
9.17 |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 215-234-0
TSCA : TSCA
RUO – Research Use Only