missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ytterbium ingot, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 00040706

 View more versions of this product

Catalog No. AA0040706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
Specifications

Specifications

Ytterbium ingot
REO
MFCD00011286
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
[Yb]
173.05
CHEBI:33381
Odorless
Ingot
7440-64-4
Yb
5 g
NAWDYIZEMPQZHO-UHFFFAOYSA-N
ytterbium
23992
99.9%
Poly Bottle

Chemical Identifiers

7440-64-4
173.05
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
ytterbium
Yb
MFCD00011286
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
CHEBI:33381
[Yb]
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 231-173-2

TSCA : Yes

storageNote1 :

  • Packaged in air stable poly bottle

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only