missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

$259.41 - $778.49

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA12443FF
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
012443FF
25 x 25 mm
Each for $259.41
 
AA12443FI
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
012443FI
50 x 50 mm
Each for $778.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]

Specifications

7440-65-5
MFCD00011468
25 x 25 mm
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
99.9%
≈0.07g/25x25mm
Yttrium foil, 0.025mm (0.001 in.) thick
Y
UN3178
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Odorless
Foil
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378s

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-174-8

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only