missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium foil, 0.64mm (0.025in) thick, 99.5% (REO), Thermo Scientific Chemicals

Form: Approx. 1.8g/25 x 25mm

Supplier:  Thermo Scientific Chemicals 000616FI

 View more versions of this product

Catalog No. AA00616FI


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
Rare Earth Oxides Content
Gray foil
99.90%
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Mylar under Argon
Yttrium
7440-65-5
4.47g/cm3
Y
50 x 50 mm
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
Odorless
Foil

Chemical Identifiers

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-174-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only