missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 199182500

Catalog No. AC199182500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for ultra trace analysis
Specifications

Specifications

Yttrium(III) chloride hexahydrate
10025-94-2
99.90%
YCl3·6H2O
14, 10107
303.35
99.9%
Glass bottle
99.9%
250g
Cl36H2O
MFCD00149941
Solubility in water: soluble. Other solubilities: soluble in alcohols
303.35
Trace Metal Basis
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 233-801-0

RTECSNumber : ZG3150000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only