Learn More
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
CAS: 13494-98-9 | N3O9Y·6H2O | 383.01 g/mol
$111.34 - $798.41
Chemical Identifiers
| CAS | 13494-98-9 |
|---|---|
| Molecular Formula | N3O9Y·6H2O |
| Molecular Weight (g/mol) | 383.01 |
| MDL Number | MFCD00149940 |
| InChI Key | QBAZWXKSCUESGU-UHFFFAOYSA-N |
| Synonym | yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate |
| PubChem CID | 166833 |
| IUPAC Name | yttrium(3+);trinitrate;hexahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC218260250
|
Thermo Scientific Chemicals
218260250 |
25 g | Glass bottle |
Each for $111.34
|
|
||||
|
AC218261000
|
Thermo Scientific Chemicals
218261000 |
100 g | Glass bottle |
Each for $254.99
|
|
||||
|
AC218265000
|
Thermo Scientific Chemicals
218265000 |
500 g | Glass bottle |
Each for $798.41
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
| 13494-98-9 | |
| 383.01 | |
| QBAZWXKSCUESGU-UHFFFAOYSA-N | |
| 166833 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] |
| N3O9Y·6H2O | |
| MFCD00149940 | |
| yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate | |
| yttrium(3+);trinitrate;hexahydrate |
Specifications
| 13494-98-9 | |
| Adhering Crystalline Powder or Crystals | |
| 99.9% | |
| Y(NO3)3·6H2O | |
| 14,10107 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] | |
| 383.01 | |
| 383.01 | |
| Trace Metal Basis | |
| Yttrium(III) nitrate hexahydrate, 99.9% |
| White | |
| 25 g | |
| N3O9Y·6H2O | |
| MFCD00149940 | |
| yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate | |
| QBAZWXKSCUESGU-UHFFFAOYSA-N | |
| yttrium(3+);trinitrate;hexahydrate | |
| 166833 | |
| 99.9% | |
| Glass bottle |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medic
GHS Signal Word: Warning
RTECSNumber : ZG3750000
RUO – Research Use Only