missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01456218

 View more versions of this product

Catalog No. AA1456218


Explore available promotions

Description

Description

Yttrium oxalate is applied in ceramics, glass, and electronics. It is an important material for tri-bands rare Earth phosphors and yttrium-iron-garnets, which are very effective microwave filters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Yttrium(III) oxalate nonahydrate
13266-82-5
Crystalline
(REO)
Y2(C2O4)3·9H2O
UN3288
CTXJKXXQOORSKU-UHFFFAOYSA-H
oxalate;yttrium(3+);nonahydrate
131872179
99.9%
(decomposition)
White
50 g
C6H18O21Y2
MFCD00150502
Insoluble in water,moderately soluble in strong mineral acids
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.O.O.O.[Y+3].[Y+3]
604.001
604.01 (441.87 Anhydrous)

Chemical Identifiers

13266-82-5
604.001
CTXJKXXQOORSKU-UHFFFAOYSA-H
oxalate;yttrium(3+);nonahydrate
C6H18O21Y2
MFCD00150502
131872179
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.O.O.O.O.O.O.O.[Y+3].[Y+3]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only