missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Yttrium ingot/button,≈50.8mm (2.0 in.) dia. x 11mm (0.43 in.) thick, 99% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045114KF
Description
Yttrium ingot/button is used as intermediates, chemical research, laboratory chemicals, Manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
≈50.8mm (2.0 in.) dia. x 11mm (0.43 in.) thick | |
(REO) | |
MFCD00011468 | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
yttrium | |
23993 | |
99% | |
≈100g/piece |
Yttrium ingot/button | |
7440-65-5 | |
Y | |
1 Pc. | |
Insoluble in water | |
[Y] | |
88.91 | |
CHEBI:33331 | |
Odorless | |
Pieces |
Chemical Identifiers
7440-65-5 | |
88.91 | |
VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
23993 | |
yttrium |
Y | |
MFCD00011468 | |
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
CHEBI:33331 | |
[Y] |
Safety and Handling
EINECSNumber : 231-174-8
RTECSNumber : ZG2980000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only