missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Yttrium powder, -40 mesh, 99.6% (REO), Thermo Scientific Chemicals

$211.62 - $753.75

Identifiants chimiques

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA4414706
Afficher les documents
Thermo Scientific Chemicals
04414706
5 g
chaque for $211.62
 
AA4414714
Afficher les documents
Thermo Scientific Chemicals
04414714
25 g
chaque for $753.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]

Spécifications

Gray
Gray powder
Y
5 g
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
-40 Mesh
Powder
7440-65-5
99.6%
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Odorless
Yttrium, Rare Earth Oxides Content
Videos
Sécurité et manipulation

Sécurité et manipulation

Take precautionary measures against static discharges.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: II

missing translation for 'einecsNumber' : 231-174-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only