missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

$245.25 - $1070.10

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40299HF
View Documents
Thermo Scientific Chemicals
040299HF
10 mm
Each for $245.25
 
AA40299DH
View Documents
Thermo Scientific Chemicals
040299DH
25 mm
Each for $414.11
 
AA40299DJ
View Documents
Thermo Scientific Chemicals
040299DJ
50 mm
Each for $716.27
 
AA40299DM
View Documents
Thermo Scientific Chemicals
040299DM
100 mm
Each for $1,070.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]

Specifications

7440-65-5
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Odorless
≈0.14g/mm
Yttrium rod, 6.35mm (0.25 in.) dia.
Y
10 mm
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
99.9%
Mylar under Argon
Cast Rod
(metals basis excluding Ta)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-174-8

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only