missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Z-Phe-Ala-fluoromethyl ketone
Supplier: Thermo Scientific Chemicals J65795LB0
Description
Cell permeable negative control for FMK-labelled caspase inhibitors. Also an irreversible inhibitor of cathepsins B, L, and S
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5 mg | |
-30°C to -10°C | |
105637-38-5 | |
386.42 | |
Soluble in DMSO or DMF at 20mM | |
Powder | |
CC(NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1)C(=O)CF | |
386.42 |
White to off-white | |
Cell permeable negative control for FMK-labelled caspase inhibitors. Also an irreversible inhibitor of cathepsins B, L, and S | |
C21H23FN2O4 | |
Z-FA-FMK; Z-Phe-Ala-FMK | |
Z-Phe-Ala-fluoromethyl ketone | |
ASXVEBPEZMSPHB-UHFFFAOYNA-N | |
benzyl N-{1-[(4-fluoro-3-oxobutan-2-yl)carbamoyl]-2-phenylethyl}carbamate |
Chemical Identifiers
105637-38-5 | |
386.42 | |
Z-FA-FMK; Z-Phe-Ala-FMK | |
CC(NC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1)C(=O)CF |
C21H23FN2O4 | |
ASXVEBPEZMSPHB-UHFFFAOYNA-N | |
benzyl N-{1-[(4-fluoro-3-oxobutan-2-yl)carbamoyl]-2-phenylethyl}carbamate |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only