missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Zenon™ pHrodo™ iFL IgG Labeling Reagents

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex.

$1406.00 - $5280.00

Specifications

Product Type Labeling Reagent
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light
Labeling Target IgG
Label Type pHrodo™ (pH sensitive)
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Species Label or Dye Color Price Quantity  
Catalog Number Mfr. No. Quantity Species Label or Dye Color Price Quantity  
Z25609 DFS Item
View Documents
Invitrogen™
Z25609
250 μL Mouse pHrodo iFL Green Green
Each for $1,774.00
 
Z25618
View Documents
Invitrogen™
Z25618
250 μL Human pHrodo Deep Red Deep Red
Each for $1,406.00
 
Z25610 DFS Item
View Documents
Invitrogen™
Z25610
250 μL Mouse pHrodo iFL Red Red
Each for $1,774.00
 
Z25622
View Documents
Invitrogen™
Z25622
250 μL Mouse pHrodo Deep Red Deep Red
Each for $1,406.00
 
Z25611 DFS Item
View Documents
Invitrogen™
Z25611
250 μL Human pHrodo iFL Green Green
Each for $1,718.00
 
Z25612 DFS Item
View Documents
Invitrogen™
Z25612
250 μL Human pHrodo iFL Red Red
Each for $1,668.00
 
Z25613
View Documents
Invitrogen™
Z25613
1.2 mL Human pHrodo iFL Green Green
Each for $5,280.00
 
Z25614
View Documents
Invitrogen™
Z25614
1.2 mL Human pHrodo iFL Red Red
Each for $5,280.00
 
Description

Description

Streamline antibody-drug conjugate screening campaigns with Invitrogen Zenon pHrodo IgG Labeling Reagents to screen for antibody internalization. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody uptake and trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than 5 minutes. Because the pHrodo™ iFL dyes dramatically increase in fluorescence as the pH of their surroundings becomes more acidic, the antibodies coupled with the Zenon™ pHrodo™ iFL IgG Labeling Reagents provide an excellent measure of endocytic activity based on the acidification of the labeled antibodies as they are ingested by the cell.

The Zenon™ pHrodo™ iFL IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon™ labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Other advantages of the pH-sensitive Zenon™ pHrodo™ iFL IgG Labeling Reagents include:

  • Simplicity—No reagent purification is required
  • Accuracy—No wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—Labeled antibodies are typically ready to use in 10 minutes
  • Specificity—Dyes are designed for the specific detection of phagocytosis and endocytosis

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon™ antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Store in refrigerator at 2°C to 8°C.

Specifications

Specifications

Labeling Reagent
Store in refrigerator (2°C to 8°C) and protect from light
IgG
pHrodo™ (pH sensitive)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.