Learn More
Zeolite Y, hydrogen, Thermo Scientific Chemicals
$59.10 - $694.90
Chemical Identifiers
CAS | 1318-02-1 |
---|---|
Molecular Formula | Al2O5Si |
Molecular Weight (g/mol) | 162.043 |
MDL Number | MFCD00132601 |
InChI Key | HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Synonym | kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural |
PubChem CID | 9942228 |
IUPAC Name | dioxosilane;oxo(oxoalumanyloxy)alumane |
SMILES | O=[Al]O[Al]=O.O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4587214
|
Thermo Scientific Chemicals
04587214 |
25 g |
Each for $59.10
|
|
|||||
AA4587222
|
Thermo Scientific Chemicals
04587222 |
100 g |
Each for $146.29
|
|
|||||
AA4587236
|
Thermo Scientific Chemicals
04587236 |
500 g |
Each for $694.90
|
|
|||||
Description
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other productsZeolite Y is used as a cracking catalyst. In the petrochemical industry, it is used in acidic form in order to increase the yield of gasoline and diesel fuel from crude oil feedstock by cracking heavy paraffins into gasoline grade naphthas. Further, it is employed in the hydrocracking units as a platinum/palladium support to increase the aromatic content of reformulated refinery products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Chemical Identifiers
1318-02-1 | |
162.043 | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
9942228 | |
O=[Al]O[Al]=O.O=[Si]=O |
Al2O5Si | |
MFCD00132601 | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
dioxosilane;oxo(oxoalumanyloxy)alumane |
Specifications
White | |
Powder | |
Al2O5Si | |
kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
dioxosilane;oxo(oxoalumanyloxy)alumane | |
9942228 |
1318-02-1 | |
25 g | |
MFCD00132601 | |
Insoluble in water. | |
O=[Al]O[Al]=O.O=[Si]=O | |
162.043 | |
Zeolite Y, hydrogen |
Safety and Handling
EINECSNumber : 215-283-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only