Restek™ Zero-Dead-Volume Valco Internal Reducing Union DFS Item
Connect two sizes of tubing, using zero-dead-volume fittings on each end.
Supplier:  Restek™ RK20152
 View more versions of this product
Catalog No. 06710520
Explore available promotions
Description
Description
  • For 0.125 to 0.0625 in. O.D. tubing
  • Includes stainless steel ferrules
Specifications
Specifications
Union
0.75 mm
HPLC