missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc, 20 Mesh, Granular, ACS, 99.8%, Spectrum™ Chemical
$242.38 - $8896.15
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18614708
|
Spectrum Chemical Mfg Cor
Z1015125GM |
125 g | Poly Bottle |
Each for $242.38
|
|
||||
18614712
|
Spectrum Chemical Mfg Cor
Z1015500GM |
500 g | Poly Bottle |
Each for $526.47
|
|
||||
18614710
|
Spectrum Chemical Mfg Cor
Z101525KG |
2.5 kg | Poly Bottle |
Each for $1,559.15
|
|
||||
18614709
|
Spectrum Chemical Mfg Cor
Z101512KG |
12 kg | Poly Pail |
Each for $5,181.26
|
|
||||
18614711
|
Spectrum Chemical Mfg Cor
Z101545KGBL |
45 kg | Fiber Drum |
Each for $8,896.15
|
|
||||
Chemical Identifiers
7440-66-6 |
Specifications
7440-66-6 | |
125 g | |
ACS | |
Approx. 20 Mesh |
1 | |
99.8% | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.