missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc, ACS Grade, LabChem™

Supplier:  LabChem LC270601

 View more versions of this product

Catalog No. LC270601

Explore available promotions

Description

Description

Specifications

Specifications

Silver
419°C
907°C
Zn
MFCD00011291
Passes Test
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
65.38
20 Mesh
Granular
Zinc
7440-66-6
100
Zn
500 g
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
ACS
Poly Bottle

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 231-175-3

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions