Learn More
Zinc carbonate hydroxide, 58%, extra pure, powder, (53% Zn min.), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 222650010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc carbonate hydroxide | |
5263-02-5 | |
Fine Crystalline Powder | |
53% min. (Zn) (Complexometry) | |
3Zn(OH)2·2ZnCO3 | |
13, 10184 | |
UOURRHZRLGCVDA-UHFFFAOYSA-D | |
pentazinc(2+) hexahydroxide dicarbonate | |
549.01 | |
Extra Pure | |
4.39 |
(53% Zn min.) | |
White | |
1 kg | |
C2H6O12Zn5 | |
MFCD00011519 | |
Solubility in water: practically insoluble | |
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O | |
548.96 | |
58% | |
Glass bottle | |
4.3900g/mL |
Chemical Identifiers
5263-02-5 | |
548.96 | |
UOURRHZRLGCVDA-UHFFFAOYSA-D | |
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O |
C2H6O12Zn5 | |
MFCD00011519 | |
pentazinc(2+) hexahydroxide dicarbonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
EINECSNumber : 226-076-7
RUO – Research Use Only