Learn More
Zinc chloride, 99.99%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
$123.76 - $434.97
Chemical Identifiers
CAS | 7646-85-7 |
---|---|
Molecular Formula | Cl2Zn |
Molecular Weight (g/mol) | 136.29 |
MDL Number | MFCD00011295 |
InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
PubChem CID | 5727 |
ChEBI | CHEBI:49976 |
IUPAC Name | dichlorozinc |
SMILES | Cl[Zn]Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC380130050
|
Thermo Scientific Chemicals
380130050 |
5 g |
Each for $123.76
|
|
||||||
AC380130250
|
Thermo Scientific Chemicals
380130250 |
25 g |
Each for $434.97
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7646-85-7 | |
136.29 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Specifications
732.0°C | |
7646-85-7 | |
Free-flowing Small Crystals | |
99.99% | |
ZnCl2 | |
15, 10331 | |
Solubility in water: 432 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and ether, freely soluble in acetone | |
Cl[Zn]Cl | |
136.29 | |
CHEBI:49976 | |
99.99% | |
Glass Bottle packed in tin can | |
Zinc chloride |
293.0°C | |
White | |
5 g | |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.29 | |
Trace Metal Basis | |
0.95 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 231-592-
RUO – Research Use Only