missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc chloride, anhydrous, 98+%, Thermo Scientific Chemicals

Zinc chloride, >-98%, Cl2Zn, CAS Number-7646-85-7, zinkchloride, zinc ii chloride, zintrace, zinc dichloride, zinc chloride zncl2, zinc chloride, anhydrous, zine dichloride, zinc chloride fume, zinc chloride, zinc butter, 500g, 732 deg.C, CHEBI:49976, 2.91

Supplier:  Thermo Scientific Chemicals A1628136

 View more versions of this product

Catalog No. AAA1628136


Explore available promotions

Description

Description

Zinc chloride is used as a catalyst, dehydrating and condensing agent, soldering flux, and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is likewise employed as a mordant in printing and dyeing materials and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in organic synthesis, for example, in the Friedel-Crafts acylatoin, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chloride is used as a catalyst, dehydrating and condensing agent, soldering flux, and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is likewise employed as a mordant in printing and dyeing materials and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in organic synthesis, for example, in the Friedel-Crafts acylatoin, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

Solubility
Soluble in water, dilute HCl, alcohol, glycerol, acetone and ether.

Notes
Hygroscopic.
Specifications

Specifications

732°C
290°C
7646-85-7
Cl2Zn
MFCD00011295
14,10132
Soluble in water,dilute HCl,alcohol,glycerol,acetone and ether.
Cl[Zn]Cl
136.28
CHEBI:49976
≥98%
Hygroscopic
Zinc chloride
anhydrous
500 g
ZnCl2
UN2331
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
Odorless
2.91

Chemical Identifiers

7646-85-7
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc
Cl2Zn
MFCD00011295
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
CHEBI:49976
Cl[Zn]Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ZINC CHLORIDE, ANHYDROUS

EINECSNumber : 231-592-0

RTECSNumber : ZH1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only