missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc fluoride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 363912500

 View more versions of this product

Catalog No. AC363912500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1500.0°C
872.0°C to 910.0°C
7783-49-5
Crystals or Powder
98%
ZnF2
06,514
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
BHHYHSUAOQUXJK-UHFFFAOYSA-L
zinc;difluoride
10129912
98%
Zinc fluoride
98%
Brown to White
250 g
F2Zn
MFCD00011298
15, 10335
Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl, hno3 and nh4oh, insoluble in alcohol
[F-].[F-].[Zn+2]
103.38
103.38
Plastic bottle

Chemical Identifiers

7783-49-5
103.38
BHHYHSUAOQUXJK-UHFFFAOYSA-L
10129912
[F-].[F-].[Zn+2]
F2Zn
MFCD00011298
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
zinc;difluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger

EINECSNumber : 232-001-9

RTECSNumber : ZH3500000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only