missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc granules, ACS, -20 mesh, 99.8% min, Thermo Scientific Chemicals
Zinc Granules; ACS; 99.8%; Alfa Aesar; -20 Mesh
Supplier: Thermo Scientific Chemicals 03660222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc | |
7440-66-6 | |
Zn | |
100 g | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
zinc | |
23994 | |
ACS Reagent | |
Granular |
For analysis ACS | |
99.8% | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
[Zn] | |
65.38 | |
CHEBI:30185 | |
-20 Mesh |
Chemical Identifiers
7440-66-6 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
23994 | |
zinc |
Zn | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
CHEBI:30185 | |
[Zn] |
Safety and Handling
EINECSNumber : 231-175-3
RTECSNumber : ZG8600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only