missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc(II) acetylacetonate, ca. 25% Zn, Thermo Scientific Chemicals

$45.73 - $147.16

Chemical Identifiers

CAS 14024-63-6
Molecular Formula C10H14O4Zn
Molecular Weight (g/mol) 263.60
MDL Number MFCD00000035
InChI Key CYDXJXDAFPJUQE-FDGPNNRMSA-L
Synonym zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
PubChem CID 131675103
SMILES [Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC388770050
SDS View Documents
Thermo Scientific Chemicals
388770050
5 g Glass bottle
Each for $45.73
 
AC388771000
SDS View Documents
Thermo Scientific Chemicals
388771000
100 g Glass bottle
Each for $147.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14024-63-6
263.60
CYDXJXDAFPJUQE-FDGPNNRMSA-L
131675103
C10H14O4Zn
MFCD00000035
zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

129.0°C to 131.0°C (10.0mmHg)
White to Ivory
Powder
C10H14O4Zn
zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
CYDXJXDAFPJUQE-FDGPNNRMSA-L
zinc;(Z)-4-oxopent-2-en-2-olate;hydrate
131675103
∼25% (Zn)
Glass bottle
124.0°C to 126.0°C
14024-63-6
5 g
MFCD00000035
Solubility in water: 6.9g/L (20°C)
[Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
263.60
263.59
Authentic
Zinc(II) acetylacetonate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-860-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only