missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Metal, 20 Mesh, Granular, Spectrum™ Chemical
$404.03 - $13033.23
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18614755
|
Spectrum Chemical Mfg Cor
Z1019500GM |
500 g | Fiber Drum |
Each for $404.03
|
|
||||
|
18614752
|
Spectrum Chemical Mfg Cor
Z1019100LBBL |
100 lb. | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18614753
|
Spectrum Chemical Mfg Cor
Z101925KG |
2.5 kg | Fiber Drum |
Each for $1,344.00
|
|
||||
|
18614754
|
Spectrum Chemical Mfg Cor
Z101912KG |
12 kg | Fiber Drum |
Each for $3,504.30
|
|
||||
Chemical Identifiers
| 7440-66-6 | |
| 65.39 |
Specifications
| 7440-66-6 | |
| 500 g | |
| Approx. 20 Mesh | |
| 65.39 | |
| Zinc Metal |
| 1 | |
| Fiber Drum | |
| Granular | |
| Ungraded |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.