missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01462930

 View more versions of this product

Catalog No. AA1462930


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc mossy
(metals basis)
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Odorless
7440-66-6
Zn
250 g
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
99%
2.5cm (0.98 in.) & down

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

RTECSNumber : ZG8600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only