missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc N-Ethyl-N-phenyldithiocarbamate 98.0+%, TCI America™

Supplier:  TCI America E0163500G

 View more versions of this product

Catalog No. E0163500G


Explore available promotions

Description

Description

Specifications

Specifications

Zinc N-Ethyl-N-phenyldithiocarbamate
206°C
C18H20N2S4Zn
500 g
KMNUDJAXRXUZQS-UHFFFAOYSA-L
zinc;N-ethyl-N-phenylcarbamodithioate
6093375
≥98.0% (T)
14634-93-6
White-Yellow
MFCD00059686
N-Ethyl-N-phenyldithiocarbamic Acid Zinc Salt
CCN(C1=CC=CC=C1)C(=S)[S-].CCN(C1=CC=CC=C1)C(=S)[S-].[Zn+2]
457.992
457.99
Crystalline Powder

Chemical Identifiers

14634-93-6
457.992
KMNUDJAXRXUZQS-UHFFFAOYSA-L
6093375
CCN(C1=CC=CC=C1)C(=S)[S-].CCN(C1=CC=CC=C1)C(=S)[S-].[Zn+2]
C18H20N2S4Zn
MFCD00059686
N-Ethyl-N-phenyldithiocarbamic Acid Zinc Salt
zinc;N-ethyl-N-phenylcarbamodithioate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2186

RTECSNumber : ZH0890000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions