missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041312A1

 View more versions of this product

Catalog No. AA41312A1


Explore available promotions

Description

Description

Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc neodecanoate
Brown
C20H40O4Zn
1 kg
Immiscible with water.
CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]
409.916
407.89
27253-29-8
102°C (216°F)
MFCD00014384
zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
VADFQXONMAETJP-UHFFFAOYSA-N
7,7-dimethyloctanoic acid;zinc
117064711
Liquid

Chemical Identifiers

27253-29-8
409.916
VADFQXONMAETJP-UHFFFAOYSA-N
117064711
CC(C)(C)CCCCCC(=O)O.CC(C)(C)CCCCCC(=O)O.[Zn]
C20H40O4Zn
MFCD00014384
zinc neodecanoate, zinc neodecanoate, zn, 7,7-dimethyloctanoyl oxy zincio 7,7-dimethyloctanoate
7,7-dimethyloctanoic acid;zinc
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 248-370-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only