missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, 99% (metals basis)

CAS: 10196-18-6 | H12N2O12Zn | 297.48 g/mol

$133.64 - $1,029.18

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
IUPAC Name zinc(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1231336
View Documents
Thermo Scientific Chemicals
01231336
500 g N/A
Only null left
 
AA12313A3
View Documents
Thermo Scientific Chemicals
012313A3
2 kg N/A
Only null left
 
AA12313A9
View Documents
Thermo Scientific Chemicals
012313A9
10 kg N/A
Only null left
 
Description

Description

Zinc nitrate hexahydrate is used in the synthesis of coordination polymers. It is utilized in the preparation of zinc oxide based nanowires. It is also used as a mordant in dyeing. It is involved in the preparation of a new microporous, organically templated zinc phosphate, which finds application as catalysts and other applications involving microporosity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10196-18-6
297.48
JGPSMWXKRPZZRG-UHFFFAOYSA-N
15865313
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
H12N2O12Zn
MFCD00149889
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
zinc(2+) hexahydrate dinitrate

Specifications

∽36°C
Crystalline
99% (metals basis)
MFCD00149889
Soluble in water,alcoholSoluble in water and ethanol.
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
297.48
297.47 (189.38 Anhydrous)
2.07 g/mL
10196-18-6
500 g
H12N2O12Zn
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc(2+) hexahydrate dinitrate
15865313
Hygroscopic
Zinc nitrate hexahydrate
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P370+P378q-P501c

H272-H302-H315-H319-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-943-8

RTECSNumber : ZH4775000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only