missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc oxide, 99.5+%
CAS: 1314-13-2 | OZn | 81.38 g/mol
$123.12 - $123.12
Chemical Identifiers
| CAS | 1314-13-2 |
|---|---|
| Molecular Formula | OZn |
| Molecular Weight (g/mol) | 81.38 |
| MDL Number | MFCD00011300 |
| InChI Key | XLOMVQKBTHCTTD-UHFFFAOYSA-N |
| Synonym | zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 |
| PubChem CID | 14806 |
| ChEBI | CHEBI:36560 |
| IUPAC Name | oxozinc |
| SMILES | O=[Zn] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC194890010
|
Thermo Scientific Chemicals
194890010 |
1 kg | Plastic bottle |
Each for $123.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1314-13-2 | |
| 81.38 | |
| XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
| 14806 | |
| oxozinc |
| OZn | |
| MFCD00011300 | |
| zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
| CHEBI:36560 | |
| O=[Zn] |
Specifications
| 1975.0°C | |
| White or White-Yellow | |
| 1 kg | |
| OZn | |
| MFCD00011300 | |
| zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
| XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
| oxozinc | |
| 14806 | |
| 81.38 | |
| Plastic bottle | |
| 5.6 | |
| Zinc oxide, 99.5+% |
| 1314-13-2 | |
| Amorphous Powder | |
| 99.5% min. (Complexometry) | |
| ZnO | |
| 15,10347 | |
| Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol | |
| O=[Zn] | |
| 81.38 | |
| CHEBI:36560 | |
| ≥99.5% | |
| 1% max. (1 g,500°C) | |
| 5.6000g/mL |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 215-222-5
RTECSNumber : ZH4810000
TSCA : TSCA
RUO – Research Use Only