missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc oxide, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 422741000

 View more versions of this product

Catalog No. AC422741000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc oxide
1314-13-2
Powder
OZn
MFCD00011300
15,10347
Solubility in water: 1.6mg/l (29°C). Other solubilities: soluble in acids and alkalies,soluble in diluted acetic or mineral acids,ammoni,um carbonate,fixed alkali hydroxide solution,insoluble in alcohol
O=[Zn]
81.38
CHEBI:36560
≥99.0%
Plastic bottle
5.6000g/mL
1975.0°C
White
100 g
ZnO
01,1294; 05,622
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.38
ACS Reagent
5.6

Chemical Identifiers

1314-13-2
81.38
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 215-222-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only