missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc Peroxide, Spectrum™ Chemical
$450.71 - $16176.44
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18614795
|
Spectrum Chemical Mfg Cor
ZI110125GM |
125 g | Amber Glass Bottle |
Each for $450.71
|
|
|||||
18614796
|
Spectrum Chemical Mfg Cor
ZI110500GM |
500 g | Amber Glass Bottle |
Each for $1,576.00
|
|
|||||
18614793
|
Spectrum Chemical Mfg Cor
ZI110100LBBL |
100 lb. | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
18614794
|
Spectrum Chemical Mfg Cor
ZI11012KGBL |
12 kg | Poly Pail |
Each for $6,346.03
|
|
|||||
Chemical Identifiers
1314-22-3 |
Specifications
1314-22-3 | |
125 g | |
Amber Glass Bottle |
1 | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.