missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphate (ortho), Puratronic™, 99.995% (metals basis)

CAS: 7779-90-0 | O8P2Zn3 | 386.08 g/mol

$166.30 - $596.67

Chemical Identifiers

CAS 7779-90-0
Molecular Formula O8P2Zn3
Molecular Weight (g/mol) 386.08
MDL Number MFCD00036282
InChI Key LRXTYHSAJDENHV-UHFFFAOYSA-H
Synonym zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
PubChem CID 24519
IUPAC Name trizinc;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1301306
View Documents
Thermo Scientific Chemicals
01301306
5 g
Each for $166.30
Only null left
 
AA1301314
View Documents
Thermo Scientific Chemicals
01301314
25 g
Each for $596.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7779-90-0
386.08
LRXTYHSAJDENHV-UHFFFAOYSA-H
24519
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]
O8P2Zn3
MFCD00036282
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
trizinc;diphosphate

Specifications

7779-90-0
Powder
O8P2Zn3
MFCD00036282
zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Zn+2].[Zn+2].[Zn+2]
386.08
386.05
Puratronic
3.75 g/mL
(metals basis)
White
5 g
Zn3(PO4)2
UN3077
LRXTYHSAJDENHV-UHFFFAOYSA-H
trizinc;diphosphate
24519
99.995%
Odorless
Zinc phosphate (ortho)
Videos
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 231-944-3

RTECSNumber : TD0590000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only