missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals

$157.58 - $2029.93

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1075918
View Documents
Thermo Scientific Chemicals
01075918
50 g
Each for $157.58
 
AA1075930
View Documents
Thermo Scientific Chemicals
01075930
250 g
Each for $616.84
 
AA10759A1
View Documents
Thermo Scientific Chemicals
010759A1
1 kg
Each for $2,029.93
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]

Specifications

Gray
Gray shot
Zn
50 g
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
65.3
Shot
7440-66-6
99.999% (Metals basis)
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Odorless
Zinc, 1-5mm (0.04-0.2 in.)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

RTECSNumber : ZG8600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only