missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc sulfate, 100mM aq. soln., Thermo Scientific™

$138.05 - $138.05

Chemical Identifiers

CAS 7733-02-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60963AE
View Documents
Thermo Scientific Chemicals
J60963AE
100 mL
Each for $138.05
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7733-02-0
161.44
NWONKYPBYAMBJT-UHFFFAOYSA-L
24424
zinc;sulfate
O4SZn
MFCD00011302
zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
CHEBI:35176
[Zn++].[O-]S([O-])(=O)=O

Specifications

Colorless
Liquid
O4SZn
MFCD00011302
zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
[Zn++].[O-]S([O-])(=O)=O
161.44
CHEBI:35176
100mM aq. soln.
7733-02-0
100 mL
ZnSO4
UN3082
NWONKYPBYAMBJT-UHFFFAOYSA-L
zinc;sulfate
24424
161.43
Zinc sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'