missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc Sulfate Heptahydrate, 99.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
$90.18 - $135.17
Chemical Identifiers
CAS | 7446-20-0 |
---|---|
Molecular Formula | H14O11SZn |
Molecular Weight (g/mol) | 287.54 |
MDL Number | MFCD00149894 |
InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
PubChem CID | 62640 |
ChEBI | CHEBI:32312 |
IUPAC Name | zinc(2+) heptahydrate sulfate |
SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19145280
|
MP Biomedicals Inc
0219145280 |
100 g |
Each for $90.18
|
|
|||||
ICN19145290
|
MP Biomedicals Inc
0219145290 |
500 g |
Each for $135.17
|
|
|||||
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
zinc(2+) heptahydrate sulfate |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Specifications
7446-20-0 | |
100 g | |
H14O11SZn | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
ACS Reagent |
Crystalline | |
99.0-102.0% | |
MFCD00149894 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc(2+) heptahydrate sulfate | |
62640 | |
287.54 | |
Zinc Sulfate Heptahydrate |
Safety and Handling
Recommended Storage : Room Temperature