missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.99% (metals basis)

Used in Luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics. | CAS: 1314-98-3 | SZn | 97.44 g/mol

$53.84 - $384.84

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
SMILES [S--].[Zn++]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA4009109
View Documents
Thermo Scientific Chemicals
04009109
10 g
Each for $53.84
Only null left
 
AA4009118
View Documents
Thermo Scientific Chemicals
04009118
50 g
Each for $113.51
Only null left
 
AA4009130
View Documents
Thermo Scientific Chemicals
04009130
250 g
Each for $384.84
Only null left
 
Description

Description

Used in Luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-98-3
97.44
DRDVZXDWVBGGMH-UHFFFAOYSA-N
14821
SZn
MFCD00011301
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
[S--].[Zn++]

Specifications

1700°C
1314-98-3
10 g
SZn
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
DRDVZXDWVBGGMH-UHFFFAOYSA-N
sulfanylidenezinc
14821
Odorless
Zinc sulfide
White
Powder
99.99% metal basis
MFCD00011301
soluble in acids,insoluble in water.
[S--].[Zn++]
97.44
97.43
4.1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-251-3

RTECSNumber : ZH5400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only