missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc wire, 0.25mm (0.01in) dia, Puratronic™, 99.994% (metals basis), Thermo Scientific Chemicals
Form: approx. 0.35g/m
$126.82 - $1683.32
Chemical Identifiers
CAS | 7440-66-6 |
---|---|
Molecular Formula | Zn |
Molecular Weight (g/mol) | 65.38 |
MDL Number | MFCD00011291 |
InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
Synonym | dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust |
PubChem CID | 23994 |
ChEBI | CHEBI:30185 |
IUPAC Name | zinc |
SMILES | [Zn] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA12055G2
|
Thermo Scientific Chemicals
012055G2 |
2 m |
Each for $126.82
|
|
|||||
AA12055G6
|
Thermo Scientific Chemicals
012055G6 |
10 m |
Each for $417.71
|
|
|||||
AA12055H2
|
Thermo Scientific Chemicals
012055H2 |
50 m |
Each for $1,683.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-66-6 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
23994 | |
zinc |
Zn | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
CHEBI:30185 | |
[Zn] |
Specifications
Gray | |
Gray wire | |
99.994% (Metals basis) | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
[Zn] | |
65.38 | |
CHEBI:30185 | |
Wire |
7440-66-6 | |
7.14g/cm3 | |
Zn | |
2 m | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
zinc | |
23994 | |
Odorless | |
Zinc |
Safety and Handling
EINECSNumber : 231-175-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only