missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 042704G9

Catalog No. AA42704G9


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7440-66-6
7.14g/cm3
Zn
25 m
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Odorless
Zinc
Gray wire
99.95% (Metals basis)
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Wire

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only