missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zincon monosodium salt, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2425614

 View more versions of this product

Catalog No. AAB2425614


Explore available promotions

Description

Description

Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.

Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Zincon monosodium salt
∼203°C (decomposition)
MFCD00064385
3872699
IABRINWJUBAIIE-JJECXDOKSA-N
2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
131856391
62625-22-3
C20H16N4NaO6S
25 g
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]
463.42
462.42

Chemical Identifiers

62625-22-3
463.42
IABRINWJUBAIIE-JJECXDOKSA-N
2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
C20H16N4NaO6S
MFCD00064385
131856391
C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 263-651-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only