missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 086108A1

 View more versions of this product

Catalog No. AA86108A1


Explore available promotions

Description

Description

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds an application as rubber additive and fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium Dichloride Oxide
15461-27-5
1 kg
Cl2OZr
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
IPCAPQRVQMIMAN-UHFFFAOYSA-L
oxozirconium;octahydrate;dihydrochloride
159678
Hygroscopic
400°C (decomposition)
Lump
99.9%, metal basis
MFCD00149898
Soluble in water and alcohol.
Cl[Zr](Cl)=O
178.12
178.13 (Anhydrous)
1.91

Chemical Identifiers

15461-27-5
178.12
IPCAPQRVQMIMAN-UHFFFAOYSA-L
159678
Cl2OZr
MFCD00149898
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
Cl[Zr](Cl)=O
Videos
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8; Packaging Group: II

EINECSNumber : 231-717-9

RTECSNumber : ZH7250000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only