missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium dichloride oxide octahydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1234222

 View more versions of this product

Catalog No. AAA1234222


Explore available promotions

Description

Description

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic, moisture sensitive and incompatible with strong oxidizing agents.
Specifications

Specifications

Zirconium dichloride oxide octahydrate
13520-92-8
Cl2H18O9Zr
UN3260
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
FUBACIUATZGHAC-UHFFFAOYSA-N
oxozirconium;octahydrate;dihydrochloride
159678
98%
Hygroscopic
150°C (decomposition)
100 g
MFCD00149898
14,10184
Soluble in water and alcohol.
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
324.26
322.25 (178.13 Anhydrous)
Acidic
1.91

Chemical Identifiers

13520-92-8
324.26
FUBACIUATZGHAC-UHFFFAOYSA-N
159678
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Cl2H18O9Zr
MFCD00149898
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
oxozirconium;octahydrate;dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-717-9

RTECSNumber : ZH7250000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only