missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.

Supplier:  Thermo Scientific Chemicals 04322414

 View more versions of this product

Catalog No. AA4322414


Explore available promotions

Description

Description

Zirconium dinitrate oxide hydrate is used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium dinitrate oxide hydrate
Crystalline
99.9% (metals basis)
MFCD00149896
Soluble in alcohol.
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
231.23
231.23 (Anhydrous)
14985-18-3
25 g
N2O7Zr
zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
JWBWFTANBHYHBC-UHFFFAOYSA-N
(nitrooxy)(oxo)zirconio nitrate
16211674

Chemical Identifiers

14985-18-3
231.23
JWBWFTANBHYHBC-UHFFFAOYSA-N
16211674
N2O7Zr
MFCD00149896
zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314

DOTInformation : Hazard Class: 5.1; Packaging Group: III

EINECSNumber : 237-529-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only