Learn More
Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 206415000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) chloride | |
98% | |
White | |
500 g | |
Cl4Zr | |
MFCD00011306 | |
13,16 | |
Zirconium tetrachloride | |
233.03 | |
98% | |
2.8 |
437.0°C | |
10026-11-6 | |
Fine Powder | |
98% | |
ZrCl4 | |
01,1295 | |
15,10374 | |
Solubility in water: reacts. Other solubilities: forming hcl and zro2, soluble in alcohol and ether | |
233.03 | |
Glass bottle |
Chemical Identifiers
10026-11-6 | |
233.03 | |
Zirconium tetrachloride |
Cl4Zr | |
MFCD00011306 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Reacts violently with water.
Corrosive to the respiratory tract.
GHS P Statement
Wear respiratory protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
C
GHS Signal Word: Danger
EINECSNumber : 233-058-2
RTECSNumber : ZH7175000
TSCA : TSCA
RUO – Research Use Only