Learn More
Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals
$89.12 - $449.38
Chemical Identifiers
CAS | 10026-11-6 |
---|---|
Molecular Formula | Cl4Zr |
Molecular Weight (g/mol) | 233.03 |
MDL Number | MFCD00011306 |
Synonym | Zirconium tetrachloride |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC206410050
|
Thermo Scientific Chemicals
206410050 |
5 g | Glass bottle |
N/A
|
|
||||
AC206411000
|
Thermo Scientific Chemicals
206411000 |
100 g | Glass bottle |
N/A
|
|
||||
AC206415000
|
Thermo Scientific Chemicals
206415000 |
500 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-11-6 | |
233.03 | |
Zirconium tetrachloride |
Cl4Zr | |
MFCD00011306 |
Specifications
437.0°C | |
White | |
5 g | |
Cl4Zr | |
MFCD00011306 | |
13,16 | |
Zirconium tetrachloride | |
233.03 | |
98% | |
2.8 |
10026-11-6 | |
Fine Powder | |
98% | |
ZrCl4 | |
01,1295 | |
15,10374 | |
Solubility in water: reacts. Other solubilities: forming hcl and zro2, soluble in alcohol and ether | |
233.03 | |
Glass bottle | |
Zirconium(IV) chloride, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Reacts violently with water.
Corrosive to the respiratory tract.
GHS P Statement
Wear respiratory protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
C
GHS Signal Word: Danger
EINECSNumber : 233-058-2
RTECSNumber : ZH7175000
TSCA : TSCA
RUO – Research Use Only