missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium(IV) isopropoxide isopropanol complex, Thermo Scientific Chemicals

$195.40 - $741.42

Chemical Identifiers

CAS 14717-56-7
Molecular Formula C15H36O5Zr
Molecular Weight (g/mol) 387.67
MDL Number MFCD00075157,MFCD00078045
InChI Key NIJDLRJGRCHJDB-UHFFFAOYSA-N
Synonym isopropanoltetrakis isopropanolato zirconium
PubChem CID 131848446
IUPAC Name propan-2-ol;propan-2-olate;zirconium(4+)
SMILES [Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8611809
View Documents
Thermo Scientific Chemicals
08611809
10 g
Each for $195.40
 
AA8611818
View Documents
Thermo Scientific Chemicals
08611818
50 g
Each for $741.42
 
Description

Description

Zirconium(IV) isopropoxide isopropanol is a suitable precursor for the synthesis of olefin-functionalized zirconium containing monomers. It is used in the manufacture of hybrid zirconium containing polymers. It is also used as a source of zirconia, a well known material having high mechanical strength, chemical durability and alkali resistance. It is also used for the chemical protection of metal substrates from acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14717-56-7
387.67
NIJDLRJGRCHJDB-UHFFFAOYSA-N
131848446
[Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
C15H36O5Zr
MFCD00075157,MFCD00078045
isopropanoltetrakis isopropanolato zirconium
propan-2-ol;propan-2-olate;zirconium(4+)

Specifications

(decomposition)
Crystalline
C15H36O5Zr
UN3181
Soluble in hot isopropanol,warm hexane and toluene; Tends to precipitate over time.
[Zr+4].CC(C)O.CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
387.67
387.67
Zirconium (IV) isopropoxide isopropanol complex
14717-56-7
10 g
MFCD00075157,MFCD00078045
isopropanoltetrakis isopropanolato zirconium
NIJDLRJGRCHJDB-UHFFFAOYSA-N
propan-2-ol;propan-2-olate;zirconium(4+)
131848446
Moisture sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

EINECSNumber : 218-522-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only