Learn More
Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf), Thermo Scientific Chemicals
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.
$82.58 - $546.75
Chemical Identifiers
CAS | 1314-23-4 |
---|---|
Molecular Formula | O2Zr |
Molecular Weight (g/mol) | 123.22 |
MDL Number | MFCD00011310 MFCD01868884 |
InChI Key | MCMNRKCIXSYSNV-UHFFFAOYSA-N |
Synonym | zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler |
PubChem CID | 62395 |
IUPAC Name | dioxozirconium |
SMILES | O=[Zr]=O |
Description
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-23-4 | |
123.22 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
62395 | |
O=[Zr]=O |
O2Zr | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
dioxozirconium |
Specifications
≈5000°C | |
1314-23-4 | |
50 g | |
MFCD00011310 MFCD01868884 | |
Insoluble in water. soluble in HF,and hot Sulphuric acid. | |
O=[Zr]=O | |
123.22 | |
123.22 | |
Plastic bottle |
∽2700°C | |
Powder | |
O2Zr | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
dioxozirconium | |
62395 | |
99.4% | |
Zirconium(IV) oxide |
Safety and Handling
EINECSNumber : 215-227-2
RTECSNumber : ZH8800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only