Learn More
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 427938000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) propoxide | |
23519-77-9,71-23-8 | |
1.0440g/mL | |
27.0 to 29% (ZrO2) | |
C12H28O4Zr | |
MFCD00015307 | |
1.044 | |
XPGAWFIWCWKDDL-UHFFFAOYSA-N | |
propan-1-olate;zirconium(4+) | |
90139 | |
ca. 70% |
ca. 70% | |
Amber to Colorless | |
23°C | |
AcroSeal™ Glass Bottle | |
Zr(OCH2CH2CH3)4 | |
800 mL | |
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt | |
CCCO[Zr](OCCC)(OCCC)OCCC | |
327.58 | |
327.57 | |
Slightly Viscous Liquid |
Chemical Identifiers
23519-77-9 | |
327.58 | |
XPGAWFIWCWKDDL-UHFFFAOYSA-N | |
90139 |
C12H28O4Zr | |
MFCD00015307 | |
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt | |
CCCO[Zr](OCCC)(OCCC)OCCC |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 245-711-9
RUO – Research Use Only