missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium Oxide, Approx. 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor Z110212KG

 View more versions of this product

Catalog No. 18614827


Explore available promotions

Description

Description

Specifications

Specifications

1314-23-4
12 kg
Reagent
1
∼99%
Poly Pail

Chemical Identifiers

1314-23-4
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.