missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zobell's Solution (ORP Standard, 220mV), LabChem™
Supplier: LabChem LC273101
Specifications
Zobell's Solution | |
ORP Standard, 220mV | |
500 mL | |
13746-66-2,7447-40-7,14459-95-1,7732-18-5 | |
K3Fe(CN)66 | |
Certified | |
1g/mL | |
Soluble in water |
220mV ± 10mV | |
Amber Poly | |
99,0.75,0.14,0.11 | |
Yellow | |
329.26 | |
Passes Test | |
1g/mL | |
Liquid |