missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zofenopril calcium

Supplier:  Thermo Scientific Chemicals 466565000

 View more versions of this product

Catalog No. AC466565000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zofenopril calcium
500 mg
249.0°C to 251.0°C
448.58
[Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1
897.16
81938-43-4
Solubility aq. soln.: .. Other solubilities: soluble in dmso, insoluble in methanol
0·5 Ca
NSYUKKYYVFVMST-LETVYOFWSA-L
calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)

Chemical Identifiers

81938-43-4
0·5 Ca
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Very toxic to aquatic life with long lasting effects.

GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only