missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zonisamide 98.0+%, TCI America™

Supplier:  TCI America Z0026200MG

 View more versions of this product

Catalog No. Z0026200MG


Explore available promotions

Description

Description

Specifications

Specifications

Zonisamide
165°C
C8H8N2O3S
200 mg
UBQNRHZMVUUOMG-UHFFFAOYSA-N
(1,2-benzoxazol-3-yl)methanesulfonamide
5734
212.22
Crystalline Powder
68291-97-4
White-Yellow
MFCD00865316
zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran
NS(=O)(=O)CC1=NOC2=CC=CC=C12
212.22
CHEBI:10127
≥98.0% (HPLC,N)

Chemical Identifiers

68291-97-4
212.22
UBQNRHZMVUUOMG-UHFFFAOYSA-N
5734
(1,2-benzoxazol-3-yl)methanesulfonamide
C8H8N2O3S
MFCD00865316
zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran
CHEBI:10127
NS(=O)(=O)CC1=NOC2=CC=CC=C12
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DE4930000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions